missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylglutaconic acid, Mixture of E and Z Isomers, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 5746-90-7 |
| Linear Formula | C6H8O4 |
| Quantity | 250 mg |
| Molecular Weight (g/mol) | 144.13 |
| Formula Weight | 144.13 |
| Percent Purity | ≥98.0% (HPLC) |
| Chemical Name or Material | 3-Methylglutaconic Acid |