missing translation for 'onlineSavingsMsg'
Learn More

3-Methylfuran (stabilized with HQ) 98.0+%, TCI America™

Catalog No. M09391G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09391G 1 g
M09395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09391G Supplier TCI America Supplier No. M09391G

Chemical Identifiers

CAS 930-27-8
Molecular Formula C5H6O
Molecular Weight (g/mol) 82.102
MDL Number MFCD00060134
InChI Key KJRRQXYWFQKJIP-UHFFFAOYSA-N
Synonym furan, 3-methyl, 3-methylfurane, 3-methyl furan, 3-methyl-furan, 3-methyl-fura, 3-methylfuran, pubchem19993, 3-methylfuran-methyl-d3, acmc-20an04, 3-methylfuran 1gr
PubChem CID 13587
IUPAC Name 3-methylfuran
SMILES CC1=COC=C1

Specifications

Color Yellow
Boiling Point 66°C
Quantity 1 g
UN Number 1993
Formula Weight 82.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylfuran (stabilized with HQ)
RTECSNumber LU2630000
TSCA No
Recommended Storage Refrigerator