missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylflavone-8-carboxylic Acid 98.0+%, TCI America™
$68.97 - $205.05
Chemical Identifiers
| CAS | 3468-01-7 |
|---|---|
| Molecular Formula | C17H12O4 |
| Molecular Weight (g/mol) | 280.28 |
| MDL Number | MFCD00040998 |
| InChI Key | KMMBBZOSQNLLMN-UHFFFAOYSA-N |
| Synonym | 3-methylflavone-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-4h-chromene-8-carboxylic acid, 8-carboxy-3-methylflavone, 4h-1-benzopyran-8-carboxylicacid, 3-methyl-4-oxo-2-phenyl, unii-o8rcm70c48, 4h-1-benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl, 3-methyl-4-oxo-2-phenyl-4h-1-benzopyran-8-carboxylic acid, enamine_005545, 3-methylflavone-8-carboxylicacid, acmc-20a6bq |
| PubChem CID | 77016 |
| ChEBI | CHEBI:61237 |
| IUPAC Name | 3-methyl-4-oxo-2-phenylchromene-8-carboxylic acid |
| SMILES | CC1=C(OC2=C(C1=O)C=CC=C2C(=O)O)C3=CC=CC=C3 |
Chemical Identifiers
| 3468-01-7 | |
| 280.28 | |
| KMMBBZOSQNLLMN-UHFFFAOYSA-N | |
| 77016 | |
| 3-methyl-4-oxo-2-phenylchromene-8-carboxylic acid |
| C17H12O4 | |
| MFCD00040998 | |
| 3-methylflavone-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-4h-chromene-8-carboxylic acid, 8-carboxy-3-methylflavone, 4h-1-benzopyran-8-carboxylicacid, 3-methyl-4-oxo-2-phenyl, unii-o8rcm70c48, 4h-1-benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl, 3-methyl-4-oxo-2-phenyl-4h-1-benzopyran-8-carboxylic acid, enamine_005545, 3-methylflavone-8-carboxylicacid, acmc-20a6bq | |
| CHEBI:61237 | |
| CC1=C(OC2=C(C1=O)C=CC=C2C(=O)O)C3=CC=CC=C3 |
Specifications
| 3468-01-7 | |
| White-Yellow | |
| MFCD00040998 | |
| 3-methylflavone-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-4h-chromene-8-carboxylic acid, 8-carboxy-3-methylflavone, 4h-1-benzopyran-8-carboxylicacid, 3-methyl-4-oxo-2-phenyl, unii-o8rcm70c48, 4h-1-benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl, 3-methyl-4-oxo-2-phenyl-4h-1-benzopyran-8-carboxylic acid, enamine_005545, 3-methylflavone-8-carboxylicacid, acmc-20a6bq | |
| CC1=C(OC2=C(C1=O)C=CC=C2C(=O)O)C3=CC=CC=C3 | |
| 280.28 | |
| CHEBI:61237 | |
| ≥98.0% (HPLC,T) | |
| 3-Methylflavone-8-carboxylic Acid |
| 236°C | |
| C17H12O4 | |
| 5 g | |
| KMMBBZOSQNLLMN-UHFFFAOYSA-N | |
| 3-methyl-4-oxo-2-phenylchromene-8-carboxylic acid | |
| 77016 | |
| 280.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No