missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylflavone-8-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America M237125G
Specifications
| 3-Methylflavone-8-carboxylic Acid | |
| 236°C | |
| C17H12O4 | |
| 25 g | |
| KMMBBZOSQNLLMN-UHFFFAOYSA-N | |
| 3-methyl-4-oxo-2-phenylchromene-8-carboxylic acid | |
| 77016 | |
| 280.28 | |
| Crystalline Powder |
| 3468-01-7 | |
| White-Yellow | |
| MFCD00040998 | |
| 3-methylflavone-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-4h-chromene-8-carboxylic acid, 8-carboxy-3-methylflavone, 4h-1-benzopyran-8-carboxylicacid, 3-methyl-4-oxo-2-phenyl, unii-o8rcm70c48, 4h-1-benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl, 3-methyl-4-oxo-2-phenyl-4h-1-benzopyran-8-carboxylic acid, enamine_005545, 3-methylflavone-8-carboxylicacid, acmc-20a6bq | |
| CC1=C(OC2=C(C1=O)C=CC=C2C(=O)O)C3=CC=CC=C3 | |
| 280.28 | |
| CHEBI:61237 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 3468-01-7 | |
| 280.28 | |
| KMMBBZOSQNLLMN-UHFFFAOYSA-N | |
| 77016 | |
| 3-methyl-4-oxo-2-phenylchromene-8-carboxylic acid |
| C17H12O4 | |
| MFCD00040998 | |
| 3-methylflavone-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-4h-chromene-8-carboxylic acid, 8-carboxy-3-methylflavone, 4h-1-benzopyran-8-carboxylicacid, 3-methyl-4-oxo-2-phenyl, unii-o8rcm70c48, 4h-1-benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl, 3-methyl-4-oxo-2-phenyl-4h-1-benzopyran-8-carboxylic acid, enamine_005545, 3-methylflavone-8-carboxylicacid, acmc-20a6bq | |
| CHEBI:61237 | |
| CC1=C(OC2=C(C1=O)C=CC=C2C(=O)O)C3=CC=CC=C3 |
Safety and Handling
TSCA : No