missing translation for 'onlineSavingsMsg'
Learn More

3-Methyldiphenylamine 98.0+%, TCI America™

Catalog No. M125825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M125825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M125825G Supplier TCI America Supplier No. M125825G

Chemical Identifiers

CAS 1205-64-7
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.254
MDL Number MFCD00008530
InChI Key TWPMMLHBHPYSMT-UHFFFAOYSA-N
Synonym 3-methyldiphenylamine, n-phenyl-m-toluidine, benzenamine, 3-methyl-n-phenyl, n-phenyl-3-methylaniline, 3-methyl diphenylamine, 3-methyl diphenyl amine, 3-methyl-n-phenyl-aniline, n-3-methylphenyl-aniline, 3-methylphenyl phenylamine, n-3-methylphenyl aniline
PubChem CID 14569
IUPAC Name 3-methyl-N-phenylaniline
SMILES CC1=CC(=CC=C1)NC2=CC=CC=C2

Specifications

Color Yellow
Quantity 25 g
Formula Weight 183.25
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 3-Methyldiphenylamine