missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcyclohexylamine (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. M21055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21055G 5 g
M210525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21055G Supplier TCI America Supplier No. M21055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6850-35-7
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00001494
InChI Key JYDYHSHPBDZRPU-UHFFFAOYNA-N
Synonym 1-Amino-3-methylcyclohexane
PubChem CID 110931
IUPAC Name 3-methylcyclohexan-1-amine
SMILES CC1CCCC(N)C1

Specifications

Color Colorless
Boiling Point 151°C
Quantity 5 g
UN Number 2733
Formula Weight 113.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methylcyclohexylamine (cis- and trans- mixture)