accessibility menu, dialog, popup

UserName

CAS RN 6850-35-7

CH3H2N

CAS RN 6850-35-7

IUPAC Name: 3-methylcyclohexan-1-amine
Synonyms: 3-methylcyclohexan-1-amine
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: JYDYHSHPBDZRPU-UHFFFAOYNA-N
SMILES: CC1CCCC(N)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results