missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylcyclohexanone 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 169°C |
| Quantity | 500 mL |
| UN Number | 2297 |
| Formula Weight | 112.17 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Methylcyclohexanone |
Safety and Handling
| EINECSNumber | (3)-2411 |
| RTECSNumber | GW1750100 |
| TSCA | Yes |