missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcyclohexane-1,2-dione, 98+%

Catalog No. p-7026566
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2262306 5 g
AAB2262314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2262306 Supplier Thermo Scientific Chemicals Supplier No. B2262306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3008-43-3 | C7H10O2 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3008-43-3
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00209518
InChI Key JDXJKLGWPNXSHL-UHFFFAOYNA-N
Synonym 3-methyl-1,2-cyclohexanedione, 1,2-cyclohexanedione, 3-methyl, 1-methyl-2,3-cyclohexadione, coffee dione, fema no. 3305, methylcyclohexanedione, acmc-1cmqp, dsstox_cid_27675, dsstox_rid_82493, dsstox_gsid_47675
PubChem CID 3321360
IUPAC Name 3-methylcyclohexane-1,2-dione
SMILES CC1CCCC(=O)C1=O

Specifications

Melting Point 57°C to 59°C
Boiling Point 69°C to 72°C (1 mmHg)
Quantity 5 g
Formula Weight 126.16
Percent Purity ≥98%
Chemical Name or Material 3-Methylcyclohexane-1,2-dione
EINECSNumber 221-122-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only