accessibility menu, dialog, popup

UserName

CAS RN 3008-43-3

IUPAC Name: 3-methylcyclohexane-1,2-dione
Synonyms: methylcyclohexanedione
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: JDXJKLGWPNXSHL-UHFFFAOYNA-N
SMILES: CC1CCCC(=O)C1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)