missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylcrotonic Acid 98.0+%, TCI America™
$33.56 - $285.21
Chemical Identifiers
| CAS | 541-47-9 |
|---|---|
| Molecular Formula | C5H8O2 |
| Molecular Weight (g/mol) | 100.117 |
| MDL Number | MFCD00004366 |
| InChI Key | YYPNJNDODFVZLE-UHFFFAOYSA-N |
| Synonym | 3,3-dimethylacrylic acid, senecioic acid, 3-methylcrotonic acid, 3-methyl-2-butenoic acid, 2-butenoic acid, 3-methyl, senecic acid, beta,beta-dimethylacrylic acid, crotonic acid, 3-methyl, beta,beta-dimethacrylic acid, beta-methylcrotonic acid |
| PubChem CID | 10931 |
| ChEBI | CHEBI:37127 |
| IUPAC Name | 3-methylbut-2-enoic acid |
| SMILES | CC(=CC(=O)O)C |
Chemical Identifiers
| 541-47-9 | |
| 100.117 | |
| YYPNJNDODFVZLE-UHFFFAOYSA-N | |
| 10931 | |
| 3-methylbut-2-enoic acid |
| C5H8O2 | |
| MFCD00004366 | |
| 3,3-dimethylacrylic acid, senecioic acid, 3-methylcrotonic acid, 3-methyl-2-butenoic acid, 2-butenoic acid, 3-methyl, senecic acid, beta,beta-dimethylacrylic acid, crotonic acid, 3-methyl, beta,beta-dimethacrylic acid, beta-methylcrotonic acid | |
| CHEBI:37127 | |
| CC(=CC(=O)O)C |
Specifications
| 541-47-9 | |
| White-Yellow | |
| C5H8O2 | |
| 25 g | |
| 3,3-dimethylacrylic acid, senecioic acid, 3-methylcrotonic acid, 3-methyl-2-butenoic acid, 2-butenoic acid, 3-methyl, senecic acid, beta,beta-dimethylacrylic acid, crotonic acid, 3-methyl, beta,beta-dimethacrylic acid, beta-methylcrotonic acid | |
| CC(=CC(=O)O)C | |
| 100.117 | |
| CHEBI:37127 | |
| ≥98.0% (GC,T) | |
| 3-Methylcrotonic Acid |
| 68°C | |
| 98°C | |
| MFCD00004366 | |
| 3261 | |
| YYPNJNDODFVZLE-UHFFFAOYSA-N | |
| 3-methylbut-2-enoic acid | |
| 10931 | |
| 100.12 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0609
RTECSNumber : GQ5425000
TSCA : Yes