missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylcrotonic Acid 98.0+%, TCI America™
Supplier: TCI America M0543100G
Specifications
| 3-Methylcrotonic Acid | |
| 68°C | |
| 98°C | |
| MFCD00004366 | |
| 3261 | |
| YYPNJNDODFVZLE-UHFFFAOYSA-N | |
| 3-methylbut-2-enoic acid | |
| 10931 | |
| 100.12 | |
| Crystalline Powder |
| 541-47-9 | |
| White-Yellow | |
| C5H8O2 | |
| 100 g | |
| 3,3-dimethylacrylic acid, senecioic acid, 3-methylcrotonic acid, 3-methyl-2-butenoic acid, 2-butenoic acid, 3-methyl, senecic acid, beta,beta-dimethylacrylic acid, crotonic acid, 3-methyl, beta,beta-dimethacrylic acid, beta-methylcrotonic acid | |
| CC(=CC(=O)O)C | |
| 100.117 | |
| CHEBI:37127 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 541-47-9 | |
| 100.117 | |
| YYPNJNDODFVZLE-UHFFFAOYSA-N | |
| 10931 | |
| 3-methylbut-2-enoic acid |
| C5H8O2 | |
| MFCD00004366 | |
| 3,3-dimethylacrylic acid, senecioic acid, 3-methylcrotonic acid, 3-methyl-2-butenoic acid, 2-butenoic acid, 3-methyl, senecic acid, beta,beta-dimethylacrylic acid, crotonic acid, 3-methyl, beta,beta-dimethacrylic acid, beta-methylcrotonic acid | |
| CHEBI:37127 | |
| CC(=CC(=O)O)C |
Safety and Handling
EINECSNumber : (2)-0609
RTECSNumber : GQ5425000
TSCA : Yes