missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcinnamic Acid 98.0+%, TCI America™

Catalog No. p-7132649
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12985G 5 g
M129825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12985G Supplier TCI America Supplier No. M12985G
Only null left

Chemical Identifiers

CAS 3029-79-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00016844
InChI Key JZINNAKNHHQBOS-AATRIKPKSA-N
Synonym 3-methylcinnamic acid, m-methylcinnamic acid, e-3-m-tolyl acrylic acid, 3-m-tolyl acrylic acid, 2e-3-3-methylphenyl acrylic acid, 2e-3-3-methylphenyl prop-2-enoic acid, cinnamic acid, m-methyl, 3-methylcinnamic acid, predominantly trans, 2e-3-3-methylphenyl-2-propenoic acid, 2-propenoic acid, 3-3-methylphenyl-, 2e
PubChem CID 2063448
ChEBI CHEBI:61118
IUPAC Name (E)-3-(3-methylphenyl)prop-2-enoic acid
SMILES CC1=CC=CC(=C1)C=CC(=O)O

Specifications

Melting Point 116°C
Color White
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylcinnamic Acid