accessibility menu, dialog, popup

UserName

CH3OO(E)

CAS RN 3029-79-6

IUPAC Name: (2E)-3-(3-methylphenyl)prop-2-enoate
Synonyms: (2E)-3-(3-methylphenyl)prop-2-enoate
Molecular Weight (g/mol): 161.18
Molecular Formula: C10H9O2
InChi Key: JZINNAKNHHQBOS-AATRIKPKSA-M
SMILES: CC1=CC=CC(\C=C\C([O-])=O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)