missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcatechol, 97%

Catalog No. AAA1232422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1232422 100 g
AAA1232414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1232422 Supplier Thermo Scientific Chemicals Supplier No. A1232422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 488-17-5 | C7H8O2 | 124.139 g/mol

Used in organic synthesis. Used for the synthesis of antibacterial agent, antioxidants, As a polymer inhibitor, stabilizer, flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in organic synthesis. Used for the synthesis of antibacterial agent, antioxidants, As a polymer inhibitor, stabilizer, flavors.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 488-17-5
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00016435
InChI Key PGSWEKYNAOWQDF-UHFFFAOYSA-N
Synonym 3-methylcatechol, 2,3-dihydroxytoluene, 3-methylpyrocatechol, 1,2-benzenediol, 3-methyl, 2,3-toluenediol, dihydroxytoluene, pyrocatechol, 3-methyl, catechol, 3-methyl, 1,2-dihydroxy-3-methylbenzene, 3-methyl-1,2-benzenediol
PubChem CID 340
ChEBI CHEBI:18404
IUPAC Name 3-methylbenzene-1,2-diol
SMILES CC1=C(C(=CC=C1)O)O

Specifications

Melting Point 62°C to 68°C
Boiling Point 241°C
Flash Point 140°C (284°F)
Quantity 100 g
Beilstein 774602
Solubility Information Soluble in water.
Formula Weight 124.14
Percent Purity 98%
Chemical Name or Material 3-Methylcatechol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-672-6
RTECSNumber UX1910000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only