missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylcarbamoyl)benzeneboronic acid, 98%

Catalog No. p-7035424
Change view
Click to view available options
Quantity:
250 mg
1 g
Catalog No. Quantity
AAH53141MD 250 mg
AAH5314103 1 g
2 options

Catalog No. AAH53141MD

Supplier: Thermo Scientific Chemicals H53141MD

Only null left

CAS: 832695-88-2 | C8H10BNO3 | 178.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 832695-88-2
Molecular Formula C8H10BNO3
Molecular Weight (g/mol) 178.98
MDL Number MFCD04038918
InChI Key FYFFPNFUVMBPRZ-UHFFFAOYSA-N
Synonym 3-n-methylaminocarbonyl phenylboronic acid, 3-methylcarbamoyl phenyl boronic acid, 3-methylcarbamoyl benzeneboronic acid, 3-methylcarbamoyl phenylboronic acid, 3-n-methylaminocarbonyl benzeneboronic acid, 3-borono-n-methylbenzamide, boronic acid, 3-methylamino carbonyl phenyl, acmc-209prq, ksc496e8l
PubChem CID 2773524
IUPAC Name [3-(methylcarbamoyl)phenyl]boronic acid
SMILES CNC(=O)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point 114°C to 124°C
Quantity 250 mg
Formula Weight 178.98
Percent Purity 98%
Chemical Name or Material 3-(Methylcarbamoyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only