missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Methylbutyl)benzene 98.0+%, TCI America™
Supplier: TCI America M065625ML
Specifications
| (3-Methylbutyl)benzene | |
| Yellow | |
| C11H16 | |
| 25 mL | |
| XNXIYYFOYIUJIW-UHFFFAOYSA-N | |
| 3-methylbutylbenzene | |
| 16294 | |
| ≥98.0% (GC) |
| 2049-94-7 | |
| 199°C | |
| MFCD00008948 | |
| isoamylbenzene, isopentylbenzene, 3-methylbutyl benzene, benzene, 3-methylbutyl, benzene, isopentyl, 1-phenyl-3-methylbutane, 2-methyl-4-phenylbutane, 3-methyl-1-phenylbutane, isoamylbenzol, 1-isopentylbenzene | |
| CC(C)CCC1=CC=CC=C1 | |
| 148.249 | |
| 148.25 | |
| Liquid |
Chemical Identifiers
| 2049-94-7 | |
| 148.249 | |
| XNXIYYFOYIUJIW-UHFFFAOYSA-N | |
| 16294 | |
| CC(C)CCC1=CC=CC=C1 |
| C11H16 | |
| MFCD00008948 | |
| isoamylbenzene, isopentylbenzene, 3-methylbutyl benzene, benzene, 3-methylbutyl, benzene, isopentyl, 1-phenyl-3-methylbutane, 2-methyl-4-phenylbutane, 3-methyl-1-phenylbutane, isoamylbenzol, 1-isopentylbenzene | |
| 3-methylbutylbenzene |
Safety and Handling
EINECSNumber : (3)-0022
TSCA : No