missing translation for 'onlineSavingsMsg'
Learn More

(3-Methylbutyl)benzene 98.0+%, TCI America™

Catalog No. p-7132284
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
M06565ML 5 mL
M065625ML 25 mL
2 options

Catalog No. M06565ML

Supplier: TCI America M06565ML

Only null left

Chemical Identifiers

CAS 2049-94-7
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00008948
InChI Key XNXIYYFOYIUJIW-UHFFFAOYSA-N
Synonym isoamylbenzene, isopentylbenzene, 3-methylbutyl benzene, benzene, 3-methylbutyl, benzene, isopentyl, 1-phenyl-3-methylbutane, 2-methyl-4-phenylbutane, 3-methyl-1-phenylbutane, isoamylbenzol, 1-isopentylbenzene
PubChem CID 16294
IUPAC Name 3-methylbutylbenzene
SMILES CC(C)CCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 199°C
Quantity 5 mL
Formula Weight 148.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (3-Methylbutyl)benzene
EINECSNumber (3)-0022
TSCA No