missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylbutanol, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™
3-Methylbutanol belongs to the group of high chain alcohols produced as metabolites from the degradation of amino acids via keto acids. It is a volatile flavor compound, highly present in alcoholic beverages.
$560.16 - $560.16
Specifications
| CAS | 123-51-3 |
|---|---|
| Density | 0.809 g/mL (at 20°C) |
| Physical Form | Neat |
| Linear Formula | (CH3)2 CHCH2CH2OH |
| MDL Number | MFCD00002934 |
Specifications
| 123-51-3 | |
| Neat | |
| MFCD00002934 | |
| 3-Methyl-1-butanol; Isoamyl alcohol; Isopentyl alcohol | |
| ≥99.8% (GC) |
| 0.809 g/mL (at 20°C) | |
| (CH3)2 CHCH2CH2OH | |
| 131°C to 132°C | |
| 88.15 | |
| Analytical Standard |