missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbiphenyl, 95%

Catalog No. AAL0100603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0100603 1 g
AAL0100606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0100603 Supplier Thermo Scientific Chemicals Supplier No. L0100603

CAS: 643-93-6 | C13H12 | 168.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 643-93-6
Molecular Formula C13H12
Molecular Weight (g/mol) 168.239
MDL Number MFCD00008533
InChI Key NPDIDUXTRAITDE-UHFFFAOYSA-N
Synonym 3-methylbiphenyl, 3-phenyltoluene, 3-methyl-1,1'biphenyl, 1,1'-biphenyl, 3-methyl, 3-methyl-1,1'-biphenyl, biphenyl, 3-methyl, unii-kfp6eo4j6p, kfp6eo4j6p, 1-methyl-3-phenyl-benzene, 3-methyl-biphenyl
PubChem CID 12564
IUPAC Name 1-methyl-3-phenylbenzene
SMILES CC1=CC=CC(=C1)C2=CC=CC=C2

Specifications

Melting Point 4°C to 5°C
Density 1.014
Boiling Point 272°C
Flash Point >110°C (230°F)
Refractive Index 1.602
Quantity 1 g
Beilstein 2039658
Formula Weight 168.24
Percent Purity 95%
Chemical Name or Material 3-Methylbiphenyl
EINECSNumber 211-404-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only