missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzylamine 97.0+%, TCI America™

$38.51 - $159.79

Chemical Identifiers

CAS 100-81-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008118
InChI Key RGXUCUWVGKLACF-UHFFFAOYSA-N
Synonym 3-methylbenzylamine, m-tolylmethanamine, 3-methylphenyl methanamine, benzenemethanamine, 3-methyl, m-methylbenzylamine, 1-3-methylphenyl methanamine, 3-methylbenzyl amine, alpha-amino-m-xylene, benzylamine, m-methyl, m-xylylamine
PubChem CID 66015
IUPAC Name (3-methylphenyl)methanamine
SMILES CC1=CC(=CC=C1)CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M12741ML
View Documents
TCI America
M12741ML
1 mL
Each for $38.51
Only null left
 
M12745ML
View Documents
TCI America
M12745ML
5 mL
Each for $159.79
Only null left
 
Description

Description

Specifications

Chemical Identifiers

100-81-2
121.183
RGXUCUWVGKLACF-UHFFFAOYSA-N
66015
CC1=CC(=CC=C1)CN
C8H11N
MFCD00008118
3-methylbenzylamine, m-tolylmethanamine, 3-methylphenyl methanamine, benzenemethanamine, 3-methyl, m-methylbenzylamine, 1-3-methylphenyl methanamine, 3-methylbenzyl amine, alpha-amino-m-xylene, benzylamine, m-methyl, m-xylylamine
(3-methylphenyl)methanamine

Specifications

100-81-2
C8H11N
1 mL
3-methylbenzylamine, m-tolylmethanamine, 3-methylphenyl methanamine, benzenemethanamine, 3-methyl, m-methylbenzylamine, 1-3-methylphenyl methanamine, 3-methylbenzyl amine, alpha-amino-m-xylene, benzylamine, m-methyl, m-xylylamine
CC1=CC(=CC=C1)CN
121.183
121.18
Liquid
206°C
MFCD00008118
2735
RGXUCUWVGKLACF-UHFFFAOYSA-N
(3-methylphenyl)methanamine
66015
≥97.0% (GC,T)
3-Methylbenzylamine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4081

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'