missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzylamine, 98%

Catalog No. AC155480010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155480010 1 g Glass bottle
AC155480050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155480010 Supplier Thermo Scientific Chemicals Supplier No. 155480010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 100-81-2 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-81-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008118
InChI Key RGXUCUWVGKLACF-UHFFFAOYSA-N
Synonym 3-methylbenzylamine, m-tolylmethanamine, 3-methylphenyl methanamine, benzenemethanamine, 3-methyl, m-methylbenzylamine, 1-3-methylphenyl methanamine, 3-methylbenzyl amine, alpha-amino-m-xylene, benzylamine, m-methyl, m-xylylamine
PubChem CID 66015
IUPAC Name (3-methylphenyl)methanamine
SMILES CC1=CC(=CC=C1)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9660g/mL
Boiling Point 202.0°C to 205.0°C
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5350 to 1.5370
Linear Formula CH3C6H4CH2NH2
Quantity 1 g
Beilstein 12, 1134
Specific Gravity 0.966
Formula Weight 121.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methylbenzylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 202-890-8
TSCA TSCA

RUO – Research Use Only