missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzyl chloride, 98%

Catalog No. p-7080997
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1078422 100 g
AAA1078436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1078422 Supplier Thermo Scientific Chemicals Supplier No. A1078422

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 620-19-9 | C8H9Cl | 140.61 g/mol

3-Methylbenzyl chloride is used as an organic light-emitting diode and as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylbenzyl chloride is used as an organic light-emitting diode and as an intermediate in organic synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.

Chemical Identifiers

CAS 620-19-9
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000909
InChI Key LZBOHNCMCCSTJX-UHFFFAOYSA-N
Synonym 3-methylbenzyl chloride, 1-chloromethyl-3-methylbenzene, m-xylyl chloride, 3-methylbenzylchloride, alpha-chloro-m-xylene, m-methylbenzyl chloride, 1-chloromethyl-3-methyl-benzene, m-chloromethyl toluene, 3-chloromethyl toluene, benzene, 1-chloromethyl-3-methyl
PubChem CID 12102
IUPAC Name 1-(chloromethyl)-3-methylbenzene
SMILES CC1=CC(=CC=C1)CCl

Specifications

Density 1.066
Boiling Point 195°C to 196°C
Flash Point 75°C (167°F)
Odor Pungent
Refractive Index 1.535
Quantity 100 g
UN Number UN3265
Beilstein 2040357
Merck Index 14,10091
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 140.61
Percent Purity 98%
Chemical Name or Material 3-Methylbenzyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 210-628-9
TSCA Yes

RUO – Research Use Only