Learn More
3-Methylbenzyl bromide, 97%
CAS: 620-13-3 | C8H9Br | 185.064 g/mol
$60.74 - $165.22
Chemical Identifiers
| CAS | 620-13-3 |
|---|---|
| Molecular Formula | C8H9Br |
| Molecular Weight (g/mol) | 185.064 |
| MDL Number | MFCD00000177 |
| InChI Key | FWLWTILKTABGKQ-UHFFFAOYSA-N |
| Synonym | 3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo |
| PubChem CID | 12099 |
| IUPAC Name | 1-(bromomethyl)-3-methylbenzene |
| SMILES | CC1=CC(=CC=C1)CBr |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2473106
|
Thermo Scientific Chemicals
B2473106 |
5 g |
Each for $60.74
|
|
|||||
|
AAB2473114
|
Thermo Scientific Chemicals
B2473114 |
25 g |
Each for $165.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 620-13-3 | |
| 185.064 | |
| FWLWTILKTABGKQ-UHFFFAOYSA-N | |
| 12099 | |
| CC1=CC(=CC=C1)CBr |
| C8H9Br | |
| MFCD00000177 | |
| 3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo | |
| 1-(bromomethyl)-3-methylbenzene |
Specifications
| 620-13-3 | |
| 184°C to 185°C (340 mmHg) | |
| C8H9Br | |
| MFCD00000177 | |
| UN1701 | |
| 14,10090 | |
| FWLWTILKTABGKQ-UHFFFAOYSA-N | |
| 1-(bromomethyl)-3-methylbenzene | |
| 12099 | |
| 97% |
| 1.37 | |
| 82°C (179°F) | |
| 1.569 | |
| 5 g | |
| 2076425 | |
| 3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo | |
| CC1=CC(=CC=C1)CBr | |
| 185.064 | |
| 185.07 | |
| 3-Methylbenzyl bromide |
Safety and Handling
GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312+H332-H314
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLYL BROMIDE, LIQUID
EINECSNumber : 210-625-2
RTECSNumber : CY9039200
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only