missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. M112425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M112425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M112425G Supplier TCI America Supplier No. M112425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 587-03-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004646
InChI Key JJCKHVUTVOPLBV-UHFFFAOYSA-N
Synonym 3-methylbenzyl alcohol, 3-methylphenyl methanol, m-tolyl carbinol, m-tolylmethanol, benzenemethanol, 3-methyl, unii-ttg38p37u6, m-methylbenzyl alcohol, 3-methylbenzylalcohol, m-methyl benzyl alcohol, 3-methyl-benzenemethanol
PubChem CID 11476
ChEBI CHEBI:27995
IUPAC Name (3-methylphenyl)methanol
SMILES CC1=CC(=CC=C1)CO

Specifications

Color Colorless
Boiling Point 217°C
Quantity 25 g
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylbenzyl Alcohol
RTECSNumber DA4937010
TSCA No