missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog No. AAB2087514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2087514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2087514 Supplier Thermo Scientific Chemicals Supplier No. B2087514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 587-03-1 | Molecular Formula: C8H10O | Molecular Weight: 122.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 587-03-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004646
InChI Key JJCKHVUTVOPLBV-UHFFFAOYSA-N
Synonym 3-methylbenzyl alcohol, 3-methylphenyl methanol, m-tolyl carbinol, m-tolylmethanol, benzenemethanol, 3-methyl, unii-ttg38p37u6, m-methylbenzyl alcohol, 3-methylbenzylalcohol, m-methyl benzyl alcohol, 3-methyl-benzenemethanol
PubChem CID 11476
ChEBI CHEBI:27995
IUPAC Name (3-methylphenyl)methanol
SMILES CC1=CC(=CC=C1)CO

Specifications

Density 1.013
Boiling Point 216°C to 217°C
Flash Point 105°C (221°F)
Refractive Index 1.534
Quantity 25 g
Beilstein 2324516
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 3-Methylbenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-595-3
RTECSNumber DA4937010
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only