missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzophenone 95.0+%, TCI America™

Catalog No. p-7132616
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12475G 5 g
M124725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12475G Supplier TCI America Supplier No. M12475G

Chemical Identifiers

CAS 643-65-2
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00008535
InChI Key URBLVRAVOIVZFJ-UHFFFAOYSA-N
Synonym 3-methylbenzophenone, 3-methyl-benzophenone, phenyl m-tolyl methanone, benzophenone, 3-methyl, 3-methylphenyl-phenylmethanone, methanone, 3-methylphenyl phenyl, 3-methylphenyl phenyl ketone, 3-benzoyltoluene, m-methylbenzophenone, zlchem 905
PubChem CID 69511
IUPAC Name (3-methylphenyl)(phenyl)methanone
SMILES CC1=CC=CC(=C1)C(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 316°C
Quantity 5 g
Formula Weight 196.25
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material 3-Methylbenzophenone