missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzophenone, 98+%

Catalog No. AAA1956818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1956818 50 g
AAA1956809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1956818 Supplier Thermo Scientific Chemicals Supplier No. A1956818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 643-65-2 | C14H12O | 196.25 g/mol

3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 643-65-2
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00008535
InChI Key URBLVRAVOIVZFJ-UHFFFAOYSA-N
Synonym 3-methylbenzophenone, 3-methyl-benzophenone, phenyl m-tolyl methanone, benzophenone, 3-methyl, 3-methylphenyl-phenylmethanone, methanone, 3-methylphenyl phenyl, 3-methylphenyl phenyl ketone, 3-benzoyltoluene, m-methylbenzophenone, zlchem 905
PubChem CID 69511
IUPAC Name (3-methylphenyl)-phenylmethanone
SMILES CC1=CC=CC(=C1)C(=O)C1=CC=CC=C1

Specifications

Density 1.093
Boiling Point 316°C
Flash Point >110°C (230°F)
Refractive Index 1.598
Quantity 50 g
Beilstein 2045488
Solubility Information Difficult to mix.
Formula Weight 196.25
Percent Purity 99%
Chemical Name or Material 3-Methylbenzophenone
EINECSNumber 211-401-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only