missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzoic anhydride, 97%

Catalog No. AAH3273906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3273906 5 g
AAH3273903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3273906 Supplier Thermo Scientific Chemicals Supplier No. H3273906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21436-44-2 | C16H14O3 | 254.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21436-44-2
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.285
MDL Number MFCD00048083
InChI Key DGZSQLHPEZFGSI-UHFFFAOYSA-N
Synonym 3-methylbenzoic anhydride, m-toluic anhydride, 3-methylbenzene-1-carboxylic anhydride, benzoic acid, 3-methyl-, anhydride, 3-methylbenzene-1-carboxylicanhydride, 3-methylbenzoicanhydride, bis 3-methylbenzoic acid anhydride, 3-methylbenzoyl 3-methylbenzoate, benzoic acid,3-methyl-, 1,1'-anhydride
PubChem CID 253301
IUPAC Name (3-methylbenzoyl) 3-methylbenzoate
SMILES CC1=CC=CC(=C1)C(=O)OC(=O)C2=CC(=CC=C2)C

Specifications

Melting Point 66°C to 70°C
Quantity 5 g
Formula Weight 254.29
Percent Purity 97%
Chemical Name or Material 3-Methylbenzoic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only