missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzo[b]thiophene 97.0+%, TCI America™

Catalog No. M19521G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19521G 1 g
M19525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19521G Supplier TCI America Supplier No. M19521G

Chemical Identifiers

CAS 1455-18-1
Molecular Formula C9H8S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00040243
InChI Key SEBRPHZZSLCDRQ-UHFFFAOYSA-N
Synonym 3-methylbenzo b thiophene, 3-methylbenzothiophene, 3-methylthianaphthene, benzo b thiophene, 3-methyl, 3-methylthioindene, benzothiophene, 3-methyl, 3-methyl-1-thiaindene, 3-methylbenzylthiophene, 3-methyl-benzo b thiophene
PubChem CID 73817
IUPAC Name 3-methyl-1-benzothiophene
SMILES CC1=CSC2=CC=CC=C12

Specifications

Melting Point -40°C
Color Yellow
Boiling Point 127°C
Quantity 1 g
Formula Weight 148.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylbenzo[b]thiophene