accessibility menu, dialog, popup

UserName

H3CS

CAS RN 1455-18-1

IUPAC Name: 3-methyl-1-benzothiophene
Synonyms: 3-methyl-1-benzothiophene
Molecular Weight (g/mol): 148.22
Molecular Formula: C9H8S
InChi Key: SEBRPHZZSLCDRQ-UHFFFAOYSA-N
SMILES: CC1=CSC2=CC=CC=C12
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)