missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylbenzo[b]thiophene, 98%
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Notes
participates in base-catalyzed Knoevenagel condensation reaction
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Notes
participates in base-catalyzed Knoevenagel condensation reaction
Specifications
Specifications
| Melting Point | -40°C |
| Density | 1.13 |
| Boiling Point | 72°C to 74°C (2 mmHg) |
| Flash Point | 104°C (219°F) |
| Refractive Index | 1.624 |
| Quantity | 1 g |
| Beilstein | 111282 |
| Formula Weight | 148.23 |
| Percent Purity | 98% |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| EINECSNumber | 215-934-6 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only