missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzo[b]thiophene, 98%

Catalog No. AAA1577214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1577214 25 g
AAA1577203 1 g
AAA1577206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1577214 Supplier Thermo Scientific Chemicals Supplier No. A1577214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1455-18-1 | C9H8S | 148.22 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
participates in base-catalyzed Knoevenagel condensation reaction

Chemical Identifiers

CAS 1455-18-1
Molecular Formula C9H8S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00040243
InChI Key SEBRPHZZSLCDRQ-UHFFFAOYSA-N
Synonym 3-methylbenzo b thiophene, 3-methylbenzothiophene, 3-methylthianaphthene, benzo b thiophene, 3-methyl, 3-methylthioindene, benzothiophene, 3-methyl, 3-methyl-1-thiaindene, 3-methylbenzylthiophene, 3-methyl-benzo b thiophene
PubChem CID 73817
IUPAC Name 3-methyl-1-benzothiophene
SMILES CC1=CSC2=CC=CC=C12

Specifications

Melting Point -40°C
Density 1.13
Boiling Point 72°C to 74°C (2 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.624
Quantity 25 g
Beilstein 111282
Formula Weight 148.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methylbenzo[b]thiophene
Show More Show Less
EINECSNumber 215-934-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only