missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzo[b]thiophene-2-carboxylic acid, 97%

Catalog No. p-7027682
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2462403 1 g
AAB2462406 5 g
AAB2462414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2462403 Supplier Thermo Scientific Chemicals Supplier No. B2462403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3133-78-6 | C10H8O2S | 192.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3133-78-6
Molecular Formula C10H8O2S
Molecular Weight (g/mol) 192.232
MDL Number MFCD01830305
InChI Key YVKLUKXESFJRCE-UHFFFAOYSA-N
Synonym 3-methylbenzo b thiophene-2-carboxylic acid, 3-methylbenzothiophene-2-carboxylic acid, acmc-20a6oy, methylbenzothiophene-carboxylic, 3-methylthianaphthene-2-carboxylic acid, 3-methyl 2-benzo b thiophenecarboxylic acid, benzo b thiophene-2-carboxylicacid, 3-methyl
PubChem CID 821868
IUPAC Name 3-methyl-1-benzothiophene-2-carboxylic acid
SMILES CC1=C(SC2=CC=CC=C12)C(=O)O

Specifications

Melting Point 241°C to 244°C
Quantity 1 g
Formula Weight 192.24
Percent Purity 97%
Chemical Name or Material 3-Methylbenzo[b]thiophene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only