accessibility menu, dialog, popup

UserName

CAS RN 3133-78-6

IUPAC Name: 3-methyl-1-benzothiophene-2-carboxylate
Synonyms: 3-methyl-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 191.22
Molecular Formula: C10H7O2S
InChi Key: YVKLUKXESFJRCE-UHFFFAOYSA-M
SMILES: CC1=C(SC2=CC=CC=C12)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results