missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzo[b]thiophene-2-acetic acid, 97%

Catalog No. AAA1962002
Change view
Click to view available options
Quantity:
0.5 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1962002 0.5 g
AAA1962088 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1962002 Supplier Thermo Scientific Chemicals Supplier No. A1962002
Only null left

CAS: 1505-52-8 | C11H10O2S | 206.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1505-52-8
Molecular Formula C11H10O2S
Molecular Weight (g/mol) 206.259
MDL Number MFCD00052299
InChI Key MFVMWBIORCNCNB-UHFFFAOYSA-N
Synonym 2-3-methylbenzo b thiophen-2-yl acetic acid, benzo b thiophene-2-acetic acid, 3-methyl, 3-methylbenzo b thiophene-2-acetic acid, 3-methylthianaphthene-2-acetic acid, 3-methyl-1-benzothiophen-2-yl acetic acid, 2-3-methyl-1-benzothiophen-2-yl acetic acid, maybridge1_005368, 3-methyl thianaphthene-2acetic acid, 3-methylbenzo b-thiophene-2-acetic acid, benzo b thiophene-2-aceticacid,3-methyl
PubChem CID 2779867
IUPAC Name 2-(3-methyl-1-benzothiophen-2-yl)acetic acid
SMILES CC1=C(SC2=CC=CC=C12)CC(=O)O

Specifications

Melting Point 148°C to 150°C
Quantity 0.5 g
Formula Weight 206.25
Percent Purity 97%
Chemical Name or Material 3-Methylbenzo[b]thiophene-2-acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only