accessibility menu, dialog, popup

UserName

CAS RN 1505-52-8

H3COOS

CAS RN 1505-52-8

IUPAC Name: 2-(3-methyl-1-benzothiophen-2-yl)acetate
Synonyms: 2-(3-methyl-1-benzothiophen-2-yl)acetate
Molecular Weight (g/mol): 205.25
Molecular Formula: C11H9O2S
InChi Key: MFVMWBIORCNCNB-UHFFFAOYSA-M
SMILES: CC1=C(CC([O-])=O)SC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results