missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzo[b]furan-2-carboxylic acid, 97%

Catalog No. AAB2104403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2104403 1 g
AAB2104406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2104403 Supplier Thermo Scientific Chemicals Supplier No. B2104403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24673-56-1 | C10H8O3 | 176.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24673-56-1
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00060511
InChI Key YMZTUCZCQMQFMK-UHFFFAOYSA-N
Synonym 3-methylbenzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 3-methyl, 3-methyl-2-benzofurancarboxylic acid, 3-methyl-benzofuran-2-carboxylic acid, 3-methylbenzo b furan-2-carboxylic acid, cambridge id 5100668, acmc-1cp97, 3-methylbenzofuran-2-carboxylicacid, 3-methylbenzofuran 2 carboxylic acid, 2-benzofurancarboxylicacid, 3-methyl
PubChem CID 600591
IUPAC Name 3-methyl-1-benzofuran-2-carboxylic acid
SMILES CC1=C(OC2=CC=CC=C12)C(=O)O

Specifications

Melting Point 192°C to 197°C
Quantity 1 g
Beilstein 132104
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 3-Methylbenzo[b]furan-2-carboxylic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only