accessibility menu, dialog, popup

UserName

CAS RN 24673-56-1

IUPAC Name: 3-methyl-1-benzofuran-2-carboxylate
Synonyms: 3-methyl-1-benzofuran-2-carboxylate
Molecular Weight (g/mol): 175.16
Molecular Formula: C10H7O3
InChi Key: YMZTUCZCQMQFMK-UHFFFAOYSA-M
SMILES: CC1=C(OC2=CC=CC=C12)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)