Learn More
(3-Methylaminopropyl)trimethoxysilane, 95%
CAS: 3069-25-8 | C7H19NO3Si | 193.32 g/mol
$171.23 - $519.00
Chemical Identifiers
| CAS | 3069-25-8 |
|---|---|
| Molecular Formula | C7H19NO3Si |
| Molecular Weight (g/mol) | 193.32 |
| MDL Number | MFCD00048021 |
| InChI Key | DVYVMJLSUSGYMH-UHFFFAOYSA-N |
| Synonym | n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 |
| PubChem CID | 76480 |
| IUPAC Name | N-methyl-3-trimethoxysilylpropan-1-amine |
| SMILES | CNCCC[Si](OC)(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1654114
|
Thermo Scientific Chemicals
L1654114 |
25 g |
N/A
|
|
|||||
|
AAL1654122
|
Thermo Scientific Chemicals
L1654122 |
100 g |
N/A
|
|
|||||
Description
(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 3069-25-8 | |
| 193.32 | |
| DVYVMJLSUSGYMH-UHFFFAOYSA-N | |
| 76480 | |
| CNCCC[Si](OC)(OC)OC |
| C7H19NO3Si | |
| MFCD00048021 | |
| n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 | |
| N-methyl-3-trimethoxysilylpropan-1-amine |
Specifications
| 106°C (30mmHg) | |
| 25 g | |
| MFCD00048021 | |
| 82°C (179°F) | |
| Hydrolyzes in water. | |
| CNCCC[Si](OC)(OC)OC | |
| 193.32 | |
| 193.32 | |
| 1.419 | |
| 0.978 |
| 3069-25-8 | |
| C7H19NO3Si | |
| 2072720 | |
| n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 | |
| DVYVMJLSUSGYMH-UHFFFAOYSA-N | |
| N-methyl-3-trimethoxysilylpropan-1-amine | |
| 76480 | |
| 95% | |
| Moisture sensitive | |
| (3-Methylaminopropyl)trimethoxysilane |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P280-P305+P351+P338-P310-P370+P378q-P501c
H227-H318
EINECSNumber : 221-334-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only