missing translation for 'onlineSavingsMsg'
Learn More

(3-Methylaminopropyl)trimethoxysilane, 95%

Catalog No. AAL1654114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1654114 25 g
AAL1654122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1654114 Supplier Thermo Scientific Chemicals Supplier No. L1654114

CAS: 3069-25-8 | C7H19NO3Si | 193.32 g/mol

(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 3069-25-8
Molecular Formula C7H19NO3Si
Molecular Weight (g/mol) 193.32
MDL Number MFCD00048021
InChI Key DVYVMJLSUSGYMH-UHFFFAOYSA-N
Synonym n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27
PubChem CID 76480
IUPAC Name N-methyl-3-trimethoxysilylpropan-1-amine
SMILES CNCCC[Si](OC)(OC)OC

Specifications

Boiling Point 106°C (30mmHg)
Quantity 25 g
Beilstein 2072720
Flash Point 82°C (179°F)
Solubility Information Hydrolyzes in water.
Formula Weight 193.32
Percent Purity 95%
Refractive Index 1.419
Sensitivity Moisture sensitive
Density 0.978
Chemical Name or Material (3-Methylaminopropyl)trimethoxysilane
Show More Show Less
Hazard Category H227-H318
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P280-P305+P351+P338-P310-P370+P378q-P501c
EINECSNumber 221-334-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only