missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Methylamino)propionitrile 98.0+%, TCI America™
Supplier: TCI America M2312100ML
Specifications
| 3-(Methylamino)propionitrile | |
| Yellow | |
| C4H8N2 | |
| 100 mL | |
| UNIJBMUBHBAUET-UHFFFAOYSA-N | |
| 3-(methylamino)propanenitrile | |
| 69656 | |
| ≥98.0% (GC,T) |
| 693-05-0 | |
| 81°C | |
| MFCD00001954 | |
| 3-methylamino propanenitrile, 3-methylaminopropionitrile, 3-methylamino propionitrile, propanenitrile, 3-methylamino, 2-cyanoethyl methyl amine, propionitrile, 3-methylamino, n-2-cyanoethyl methylamine, 3-n-methylamino propionitrile, 2-cyanoethyl methylamine, n-methyl-beta-alaninenitrile | |
| CNCCC#N | |
| 84.12 | |
| 84.12 | |
| Liquid |
Chemical Identifiers
| 693-05-0 | |
| 84.12 | |
| UNIJBMUBHBAUET-UHFFFAOYSA-N | |
| 69656 | |
| CNCCC#N |
| C4H8N2 | |
| MFCD00001954 | |
| 3-methylamino propanenitrile, 3-methylaminopropionitrile, 3-methylamino propionitrile, propanenitrile, 3-methylamino, 2-cyanoethyl methyl amine, propionitrile, 3-methylamino, n-2-cyanoethyl methylamine, 3-n-methylamino propionitrile, 2-cyanoethyl methylamine, n-methyl-beta-alaninenitrile | |
| 3-(methylamino)propanenitrile |
Safety and Handling
RTECSNumber : TZ4910000
TSCA : Yes