missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Methylamino)propionitrile 98.0+%, TCI America™
$214.37 - $637.51
Chemical Identifiers
| CAS | 693-05-0 |
|---|---|
| Molecular Formula | C4H8N2 |
| Molecular Weight (g/mol) | 84.12 |
| MDL Number | MFCD00001954 |
| InChI Key | UNIJBMUBHBAUET-UHFFFAOYSA-N |
| Synonym | 3-methylamino propanenitrile, 3-methylaminopropionitrile, 3-methylamino propionitrile, propanenitrile, 3-methylamino, 2-cyanoethyl methyl amine, propionitrile, 3-methylamino, n-2-cyanoethyl methylamine, 3-n-methylamino propionitrile, 2-cyanoethyl methylamine, n-methyl-beta-alaninenitrile |
| PubChem CID | 69656 |
| IUPAC Name | 3-(methylamino)propanenitrile |
| SMILES | CNCCC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M231225ML
|
TCI America
M231225ML |
25 mL |
Each for $214.37
|
|
|||||
|
M2312100ML
|
TCI America
M2312100ML |
100 mL |
Each for $637.51
|
|
|||||
Chemical Identifiers
| 693-05-0 | |
| 84.12 | |
| UNIJBMUBHBAUET-UHFFFAOYSA-N | |
| 69656 | |
| CNCCC#N |
| C4H8N2 | |
| MFCD00001954 | |
| 3-methylamino propanenitrile, 3-methylaminopropionitrile, 3-methylamino propionitrile, propanenitrile, 3-methylamino, 2-cyanoethyl methyl amine, propionitrile, 3-methylamino, n-2-cyanoethyl methylamine, 3-n-methylamino propionitrile, 2-cyanoethyl methylamine, n-methyl-beta-alaninenitrile | |
| 3-(methylamino)propanenitrile |
Specifications
| 693-05-0 | |
| 81°C | |
| MFCD00001954 | |
| 3-methylamino propanenitrile, 3-methylaminopropionitrile, 3-methylamino propionitrile, propanenitrile, 3-methylamino, 2-cyanoethyl methyl amine, propionitrile, 3-methylamino, n-2-cyanoethyl methylamine, 3-n-methylamino propionitrile, 2-cyanoethyl methylamine, n-methyl-beta-alaninenitrile | |
| CNCCC#N | |
| 84.12 | |
| 84.12 | |
| Liquid |
| Yellow | |
| C4H8N2 | |
| 25 mL | |
| UNIJBMUBHBAUET-UHFFFAOYSA-N | |
| 3-(methylamino)propanenitrile | |
| 69656 | |
| ≥98.0% (GC,T) | |
| 3-(Methylamino)propionitrile |
Safety and Handling
RTECSNumber : TZ4910000
TSCA : Yes