Learn More
3-(Methylamino)oxetane, 95%
CAS: 952182-03-5 | C4H9NO | 87.122 g/mol
Supplier: Thermo Scientific Chemicals H57425ME
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Very Soluble (307 g/L) (25°C)
Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
Specifications
| 3-(Methylamino)oxetane | |
| 40°C (104°F) | |
| MFCD08544349 | |
| UN1993 | |
| Very Soluble (307g/L) (25°C) | |
| CNC1COC1 | |
| 87.122 | |
| 87.12 |
| 952182-03-5 | |
| C4H9NO | |
| 500 mg | |
| n-methyl-3-oxetanamine, 1-methyl-3-oxetanamine, 3-methylamino oxetane, n-methyl-3-oxetaneamine, 3-methylaminooxetane, n-methyloxetane-3-amine, 3-oxetanamine,n-methyl, n-methyl-3-aminooxetane, 3-oxetanamine, n-methyl | |
| JUIXJPRSYHSLHK-UHFFFAOYSA-N | |
| N-methyloxetan-3-amine | |
| 46839993 | |
| 95% |
Chemical Identifiers
| 952182-03-5 | |
| 87.122 | |
| JUIXJPRSYHSLHK-UHFFFAOYSA-N | |
| 46839993 | |
| CNC1COC1 |
| C4H9NO | |
| MFCD08544349 | |
| n-methyl-3-oxetanamine, 1-methyl-3-oxetanamine, 3-methylamino oxetane, n-methyl-3-oxetaneamine, 3-methylaminooxetane, n-methyloxetane-3-amine, 3-oxetanamine,n-methyl, n-methyl-3-aminooxetane, 3-oxetanamine, n-methyl | |
| N-methyloxetan-3-amine |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only