missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylamino)oxetane, 95%

Catalog No. p-7038459
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57425MD 250 mg
AAH57425ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57425MD Supplier Thermo Scientific Chemicals Supplier No. H57425MD
Only null left

CAS: 952182-03-5 | C4H9NO | 87.122 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Very Soluble (307 g/L) (25°C)

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 952182-03-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD08544349
InChI Key JUIXJPRSYHSLHK-UHFFFAOYSA-N
Synonym n-methyl-3-oxetanamine, 1-methyl-3-oxetanamine, 3-methylamino oxetane, n-methyl-3-oxetaneamine, 3-methylaminooxetane, n-methyloxetane-3-amine, 3-oxetanamine,n-methyl, n-methyl-3-aminooxetane, 3-oxetanamine, n-methyl
PubChem CID 46839993
IUPAC Name N-methyloxetan-3-amine
SMILES CNC1COC1

Specifications

Flash Point 40°C (104°F)
Quantity 250 mg
UN Number UN1993
Solubility Information Very Soluble (307g/L) (25°C)
Formula Weight 87.12
Percent Purity 95%
Chemical Name or Material 3-(Methylamino)oxetane
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only