missing translation for 'onlineSavingsMsg'
Learn More

3-Methyladenine, TCI America™

Catalog No. p-7133264
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2518200MG 200 mg
M25181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2518200MG Supplier TCI America Supplier No. M2518200MG

Chemical Identifiers

CAS 5142-23-4
Molecular Formula C6H7N5
Molecular Weight (g/mol) 149.16
MDL Number MFCD00010531
InChI Key FSASIHFSFGAIJM-UHFFFAOYSA-N
Synonym 3-methyladenine, 3-methyl-3h-purin-6-amine, 6-amino-3-methylpurine, 3-ma, 3h-purin-6-amine, 3-methyl, 3-methyl-3h-adenine, n3-methyladenine, adenine, 3-methyl, 3-methyl-3h-purin-6-ylamine, 3-methyladenine 3-ma
PubChem CID 1673
ChEBI CHEBI:38635
IUPAC Name 3-methyl-3H-purin-6-amine
SMILES CN1C=NC(N)=C2N=CN=C12

Specifications

Melting Point 300°C
Color White
Quantity 200 mg
Formula Weight 149.16
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyladenine
RTECSNumber AU6520000
TSCA No