missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3-Methyladenine, 90+%

Catalog No. AC379791000
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC379791000 100 mg Glass Bottle
AC379795000 500 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC379791000 Supplier Thermo Scientific Chemicals Supplier No. 379791000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5142-23-4 | C6H7N5 | 149.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5142-23-4
Molecular Formula C6H7N5
Molecular Weight (g/mol) 149.16
MDL Number MFCD00010531
InChI Key FSASIHFSFGAIJM-UHFFFAOYSA-N
Synonym 3-methyladenine, 3-methyl-3h-purin-6-amine, 6-amino-3-methylpurine, 3-ma, 3h-purin-6-amine, 3-methyl, 3-methyl-3h-adenine, n3-methyladenine, adenine, 3-methyl, 3-methyl-3h-purin-6-ylamine, 3-methyladenine 3-ma
PubChem CID 1673
ChEBI CHEBI:38635
SMILES CN1C=NC(N)=C2N=CN=C12

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 90% min. (HPLC)
Packaging Glass Bottle
Quantity 100 mg
Beilstein 26,V,17,151
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,dmso,chloroform
IUPAC Name 3-methyl-3H-purin-6-amine
Formula Weight 149.15
Percent Purity 90+%
Physical Form Powder
Chemical Name or Material 3-Methyladenine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
EINECSNumber 225-908-6

RUO – Research Use Only