missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-5-(trifluoromethyl)pyrazole 98.0+%, TCI America™
$70.84 - $227.41
Chemical Identifiers
| CAS | 10010-93-2 |
|---|---|
| Molecular Formula | C5H5F3N2 |
| Molecular Weight (g/mol) | 150.104 |
| MDL Number | MFCD00156058 |
| InChI Key | DLCHCAYDSKIFIN-UHFFFAOYSA-N |
| Synonym | 5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole |
| PubChem CID | 139077 |
| IUPAC Name | 5-methyl-3-(trifluoromethyl)-1H-pyrazole |
| SMILES | CC1=CC(=NN1)C(F)(F)F |
Chemical Identifiers
| 10010-93-2 | |
| 150.104 | |
| DLCHCAYDSKIFIN-UHFFFAOYSA-N | |
| 139077 | |
| CC1=CC(=NN1)C(F)(F)F |
| C5H5F3N2 | |
| MFCD00156058 | |
| 5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole | |
| 5-methyl-3-(trifluoromethyl)-1H-pyrazole |
Specifications
| 10010-93-2 | |
| White | |
| MFCD00156058 | |
| 5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole | |
| CC1=CC(=NN1)C(F)(F)F | |
| 150.104 | |
| 150.10 | |
| Crystalline Powder |
| 89°C | |
| C5H5F3N2 | |
| 1 g | |
| DLCHCAYDSKIFIN-UHFFFAOYSA-N | |
| 5-methyl-3-(trifluoromethyl)-1H-pyrazole | |
| 139077 | |
| ≥98.0% (GC) | |
| 3-Methyl-5-(trifluoromethyl)pyrazole |
Safety and Handling
TSCA : No