accessibility menu, dialog, popup

UserName

CAS RN 10010-93-2

IUPAC Name: 5-methyl-3-(trifluoromethyl)-1H-pyrazole
Synonyms: 3-methyl-5-(trifluoromethyl)-2H-pyrazole
Molecular Weight (g/mol): 150.1
Molecular Formula: C5H5F3N2
InChi Key: DLCHCAYDSKIFIN-UHFFFAOYSA-N
SMILES: CC1=CC(=NN1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results