missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(trifluoromethyl)pyrazole 98.0+%, TCI America™

Catalog No. M20325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20325G 5 g
M20321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20325G Supplier TCI America Supplier No. M20325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10010-93-2
Molecular Formula C5H5F3N2
Molecular Weight (g/mol) 150.104
MDL Number MFCD00156058
InChI Key DLCHCAYDSKIFIN-UHFFFAOYSA-N
Synonym 5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole
PubChem CID 139077
IUPAC Name 5-methyl-3-(trifluoromethyl)-1H-pyrazole
SMILES CC1=CC(=NN1)C(F)(F)F

Specifications

Melting Point 89°C
Color White
Quantity 5 g
Formula Weight 150.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-5-(trifluoromethyl)pyrazole